accessibility menu, dialog, popup

UserName

CAS RN 25152-85-6

IUPAC Name: (3Z)-hex-3-en-1-yl benzoate
Synonyms: cis-3-hexenyl benzoate
Molecular Weight (g/mol): 204.27
Molecular Formula: C13H16O2
InChi Key: BCOXBEHFBZOJJZ-ARJAWSKDSA-N
SMILES: CC\C=C/CCOC(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results