accessibility menu, dialog, popup

UserName

CAS RN 25121-83-9

IUPAC Name: 3-bromothieno[3,2-b]thiophene
Synonyms: 3-bromothieno[3,2-b]thiophene
Molecular Weight (g/mol): 219.11
Molecular Formula: C6H3BrS2
InChi Key: YFWCQBIKGWTZQQ-UHFFFAOYSA-N
SMILES: BrC1=CSC2=C1SC=C2
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results