CAS RN 25081-39-4
CAS RN 25081-39-4
IUPAC Name:
methyl 3,5-dimethylbenzoate
Synonyms:
methyl 3,5-dimethylbenzoate
Molecular Weight (g/mol):
164.2
Molecular Formula:
C10H12O2
InChi Key:
PEVXENGLERTHJE-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC(C)=CC(C)=C1
1
–
1
of
1
results
1
Methyl 3,5-dimethylbenzoate, 98%
CAS: 25081-39-4 Molecular Formula: C10H12O2 Molecular Weight (g/mol): 164.204 MDL Number: MFCD00077971 InChI Key: PEVXENGLERTHJE-UHFFFAOYSA-N PubChem CID: 32786 IUPAC Name: methyl 3,5-dimethylbenzoate SMILES: CC1=CC(=CC(=C1)C(=O)OC)C