CAS RN 25069-38-9
IUPAC Name:
4-[bis(4-bromophenyl)amino]benzaldehyde
Synonyms:
4-[bis(4-bromophenyl)amino]benzaldehyde
Molecular Weight (g/mol):
431.13
Molecular Formula:
C19H13Br2NO
InChi Key:
IDCRGMNEAOWPRP-UHFFFAOYSA-N
SMILES:
BrC1=CC=C(C=C1)N(C1=CC=C(Br)C=C1)C1=CC=C(C=O)C=C1
1
–
1
of
1
results
1
4-[N,N-Bis(4-bromophenyl)amino]benzaldehyde 98.0+%, TCI America™
CAS: 25069-38-9 Molecular Formula: C19H13Br2NO Molecular Weight (g/mol): 431.127 MDL Number: MFCD12547118 InChI Key: IDCRGMNEAOWPRP-UHFFFAOYSA-N Synonym: 4,4′C-Dibromo-4′C′C-formyltriphenylamine PubChem CID: 18668904 IUPAC Name: 4-(4-bromo-N-(4-bromophenyl)anilino)benzaldehyde SMILES: C1=CC(=CC=C1C=O)N(C2=CC=C(C=C2)Br)C3=CC=C(C=C3)Br