accessibility menu, dialog, popup

UserName

CAS RN 250220-36-1

IUPAC Name: dichloro(3-phenyl-1H-inden-1-ylidene)ruthenium; bis(tricyclohexylphosphane)
Synonyms: dichloro(3-phenylinden-1-ylidene)ruthenium; bis(tricyclohexylphosphine)
Molecular Weight (g/mol): 923.09
Molecular Formula: C51H76Cl2P2Ru
InChi Key: UZAFZWUWHHFWOK-UHFFFAOYSA-L
SMILES: Cl[Ru](Cl)=C1C=C(C2=CC=CC=C12)C1=CC=CC=C1.C1CCC(CC1)P(C1CCCCC1)C1CCCCC1.C1CCC(CC1)P(C1CCCCC1)C1CCCCC1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results