accessibility menu, dialog, popup

UserName

CAS RN 24985-85-1

H3COOHNOH

CAS RN 24985-85-1

IUPAC Name: ethyl 5-hydroxy-1H-indole-2-carboxylate
Synonyms: ethyl 5-hydroxy-1H-indole-2-carboxylate
Molecular Weight (g/mol): 205.21
Molecular Formula: C11H11NO3
InChi Key: WANAXLMRGYGCPC-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC2=CC(O)=CC=C2N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results