accessibility menu, dialog, popup

UserName

CAS RN 2494-56-6

IH3COONCH3CH3H3C

CAS RN 2494-56-6

IUPAC Name: [2-(butanoyloxy)ethyl]trimethylazanium iodide
Synonyms: butyrylcholine
Molecular Weight (g/mol): 301.17
Molecular Formula: C9H20INO2
InChi Key: GALNBQVDMJRFGJ-UHFFFAOYSA-M
SMILES: [I-].CCCC(=O)OCC[N+](C)(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results