missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Butyrylcholine Iodide 99.0+%, TCI America™
Specifications
Specifications
| CAS | 2494-56-6 |
| Melting Point | 86°C |
| Color | White-Yellow |
| Molecular Formula | C9H20INO2 |
| Quantity | 1 g |
| MDL Number | MFCD00038730 |
| InChI Key | GALNBQVDMJRFGJ-UHFFFAOYSA-M |
| SMILES | CCCC(=O)OCC[N+](C)(C)C.[I-] |
| IUPAC Name | 2-butanoyloxyethyl(trimethyl)azanium;iodide |
| Molecular Weight (g/mol) | 301.168 |
| Show More |
Safety and Handling
| RTECSNumber | BP0475000 |
| TSCA | Yes |