accessibility menu, dialog, popup

UserName

CAS RN 24938-68-9

H3COCH3

CAS RN 24938-68-9

IUPAC Name: 2-methoxy-5-methyl-3-phenyl-1,1'-biphenyl
Synonyms: 2-methoxy-5-methyl-3-phenyl-1,1'-biphenyl
Molecular Weight (g/mol): 274.36
Molecular Formula: C20H18O
InChi Key: TWADJGWUKGOPFG-UHFFFAOYSA-N
SMILES: COC1=C(C=C(C)C=C1C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results