accessibility menu, dialog, popup

UserName

CAS RN 24835-08-3

IUPAC Name: hydrogen 1-(2-nitrophenyl)methanamine chloride
Synonyms: hydrogen 1-(2-nitrophenyl)methanamine chloride
Molecular Weight (g/mol): 188.61
Molecular Formula: C7H9ClN2O2
InChi Key: BASJTVIZZDEQBJ-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NCC1=CC=CC=C1[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results