accessibility menu, dialog, popup

UserName

CAS RN 2472-22-2

IUPAC Name: 6-methoxy-1,2,3,4-tetrahydronaphthalen-2-one
Synonyms: 6-methoxy-3,4-dihydro-1H-naphthalen-2-one
Molecular Weight (g/mol): 176.22
Molecular Formula: C11H12O2
InChi Key: RMRKDYNVZWKAFP-UHFFFAOYSA-N
SMILES: COC1=CC=C2CC(=O)CCC2=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Physical Form 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)