accessibility menu, dialog, popup

UserName

CAS RN 24589-77-3

IUPAC Name: hydrogen 4-hydrazinylbenzoic acid chloride
Synonyms: hydrogen 4-hydrazinylbenzoic acid chloride
Molecular Weight (g/mol): 188.61
Molecular Formula: C7H9ClN2O2
InChi Key: XHLQMKQBCHYRLC-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NNC1=CC=C(C=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results