accessibility menu, dialog, popup

UserName

OOHNSOONOO

CAS RN 245365-64-4

IUPAC Name: benzyl N-(2-nitrobenzenesulfonyl)carbamate
Synonyms: benzyl N-(2-nitrobenzenesulfonyl)carbamate
Molecular Weight (g/mol): 336.32
Molecular Formula: C14H12N2O6S
InChi Key: BFQXFHUTPNBFSR-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC=C1S(=O)(=O)NC(=O)OCC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results