CAS RN 243977-23-3
IUPAC Name:
2-{4-[(trifluoromethyl)sulfanyl]phenyl}acetic acid
Synonyms:
{4-[(trifluoromethyl)sulfanyl]phenyl}acetic acid
Molecular Weight (g/mol):
236.21
Molecular Formula:
C9H7F3O2S
InChi Key:
BFDKCZMYQOSTJG-UHFFFAOYSA-N
SMILES:
OC(=O)CC1=CC=C(SC(F)(F)F)C=C1
1
–
1
of
1
results
1
Promotions Available
4-(Trifluoromethylthio)phenylacetic acid, 98%
4-(Trifluoromethylthio)phenylacetic acid, 98%
CAS: 243977-23-3 Molecular Formula: C9H7F3O2S Molecular Weight (g/mol): 236.208 MDL Number: MFCD00236351 InChI Key: BFDKCZMYQOSTJG-UHFFFAOYSA-N Synonym: 4-trifluoromethylthio phenylacetic acid,2-4-trifluoromethyl thio phenyl acetic acid,benzeneacetic acid, 4-trifluoromethyl thio,4-trifluoromethylsulfanyl-phenyl-acetic acid,2-4-trifluoromethylthio phenyl acetic acid,4-trifluoromethyl sulfanyl phenyl acetic acid,pubchem7296,acmc-1chv0 PubChem CID: 2777902 IUPAC Name: 2-[4-(trifluoromethylsulfanyl)phenyl]acetic acid SMILES: C1=CC(=CC=C1CC(=O)O)SC(F)(F)F