accessibility menu, dialog, popup

UserName

CAS RN 2430-99-1

IUPAC Name: 2-(4-methylphenyl)-1-phenylethan-1-one
Synonyms: 2-(4-methylphenyl)-1-phenylethanone
Molecular Weight (g/mol): 210.28
Molecular Formula: C15H14O
InChi Key: WILFDKCJCDVGQX-UHFFFAOYSA-N
SMILES: CC1=CC=C(CC(=O)C2=CC=CC=C2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results