accessibility menu, dialog, popup

UserName

CH3SHOH3COH

CAS RN 24197-34-0

IUPAC Name: 4-[(4-hydroxy-3-methylphenyl)sulfanyl]-2-methylphenol
Synonyms: 4-[(4-hydroxy-3-methylphenyl)sulfanyl]-2-methylphenol
Molecular Weight (g/mol): 246.32
Molecular Formula: C14H14O2S
InChi Key: IBNFPRMKLZDANU-UHFFFAOYSA-N
SMILES: CC1=C(O)C=CC(SC2=CC(C)=C(O)C=C2)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results