accessibility menu, dialog, popup

UserName

H3COOHOHOCH3OCH3OH3C

CAS RN 241811-66-5

IUPAC Name: [3-(hydroxymethyl)-5,6-dimethoxy-5,6-dimethyl-1,4-dioxan-2-yl]methanol
Synonyms: [3-(hydroxymethyl)-5,6-dimethoxy-5,6-dimethyl-1,4-dioxan-2-yl]methanol
Molecular Weight (g/mol): 236.26
Molecular Formula: C10H20O6
InChi Key: RAIJJZVROJTPEU-UHFFFAOYNA-N
SMILES: COC1(C)OC(CO)C(CO)OC1(C)OC
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results