accessibility menu, dialog, popup

UserName

ClS

CAS RN 24165-03-5

IUPAC Name: [(chlorosulfanyl)diphenylmethyl]benzene
Synonyms: [(chlorosulfanyl)diphenylmethyl]benzene
Molecular Weight (g/mol): 310.84
Molecular Formula: C19H15ClS
InChi Key: ZTQHJLXIUQQVST-UHFFFAOYSA-N
SMILES: ClSC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results