accessibility menu, dialog, popup

UserName

HOHNN

CAS RN 2403-66-9

IUPAC Name: 3-(1H-1,3-benzodiazol-2-yl)propan-1-ol
Synonyms: 3-(1H-1,3-benzodiazol-2-yl)propan-1-ol
Molecular Weight (g/mol): 176.22
Molecular Formula: C10H12N2O
InChi Key: CQFSGSFSOWEIGO-UHFFFAOYSA-N
SMILES: OCCCC1=NC2=CC=CC=C2N1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results