accessibility menu, dialog, popup

UserName

H3COSCH3

CAS RN 2388-73-0

IUPAC Name: 1-methoxy-2-(methylsulfanyl)benzene
Synonyms: 1-methoxy-2-(methylsulfanyl)benzene
Molecular Weight (g/mol): 154.23
Molecular Formula: C8H10OS
InChi Key: YCUXDIFPTHNTSR-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1SC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results