accessibility menu, dialog, popup

UserName

CAS RN 23876-18-8

CH3NOOOHO

CAS RN 23876-18-8

IUPAC Name: 2-(2-methyl-6-nitrophenyl)acetic acid
Synonyms: (2-methyl-6-nitrophenyl)acetic acid
Molecular Weight (g/mol): 195.17
Molecular Formula: C9H9NO4
InChi Key: WFKNETIJXXWRHO-UHFFFAOYSA-N
SMILES: CC1=C(CC(O)=O)C(=CC=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results