accessibility menu, dialog, popup

UserName

CH3

CAS RN 2381-21-7

IUPAC Name: 1-methylpyrene
Synonyms: 1-methylpyrene
Molecular Weight (g/mol): 216.28
Molecular Formula: C17H12
InChi Key: KBSPJIWZDWBDGM-UHFFFAOYSA-N
SMILES: CC1=CC=C2C=CC3=CC=CC4=CC=C1C2=C34
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results