accessibility menu, dialog, popup

UserName

H3COHCH3

CAS RN 23708-56-7

IUPAC Name: undecan-6-ol
Synonyms: 6-undecanol
Molecular Weight (g/mol): 172.31
Molecular Formula: C11H24O
InChi Key: YBIXBBGRHOUVBB-UHFFFAOYSA-N
SMILES: CCCCCC(O)CCCCC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results