accessibility menu, dialog, popup

UserName

NNNBr

CAS RN 23449-08-3

IUPAC Name: 2-(4-bromophenyl)-4,6-diphenyl-1,3,5-triazine
Synonyms: 2-(4-bromophenyl)-4,6-diphenyl-1,3,5-triazine
Molecular Weight (g/mol): 388.27
Molecular Formula: C21H14BrN3
InChi Key: AYHGAQGOMUQMTR-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)C1=NC(=NC(=N1)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results