accessibility menu, dialog, popup

UserName

SS(E/Z)(E/Z)

CAS RN 23354-93-0

IUPAC Name: 2-[4-(thiophen-2-yl)buta-1,3-dien-1-yl]thiophene
Synonyms: 2-[4-(thiophen-2-yl)buta-1,3-dien-1-yl]thiophene
Molecular Weight (g/mol): 218.33
Molecular Formula: C12H10S2
InChi Key: SEIGKYBFMXSLAH-UHFFFAOYSA-N
SMILES: S1C=CC=C1C=CC=CC1=CC=CS1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results