accessibility menu, dialog, popup

UserName

CAS RN 23256-42-0

H3CONNH2NH2NOH3COH3CH3COHOHO

CAS RN 23256-42-0

IUPAC Name: 2-hydroxypropanoic acid; 5-[(3,4,5-trimethoxyphenyl)methyl]pyrimidine-2,4-diamine
Synonyms: lactic acid; trimethoprim
Molecular Weight (g/mol): 380.4
Molecular Formula: C17H24N4O6
InChi Key: IIZVTUWSIKTFKO-UHFFFAOYNA-N
SMILES: CC(O)C(O)=O.COC1=CC(CC2=CN=C(N)N=C2N)=CC(OC)=C1OC
Physical Form 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Type 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results