accessibility menu, dialog, popup

UserName

NP

CAS RN 2325-27-1

IUPAC Name: N,1,1,1-tetraphenyl-λ⁵-phosphanimine
Synonyms: triphenylphosphine phenylimide
Molecular Weight (g/mol): 353.41
Molecular Formula: C24H20NP
InChi Key: PTLOPIHJOPWUNN-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)N=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results