CAS RN 2315-97-1
IUPAC Name:
10,10'-dimethyl-[9,9'-biacridine]-10,10'-diium dinitrate
Synonyms:
lucigenin
Molecular Weight (g/mol):
510.51
Molecular Formula:
C28H22N4O6
InChi Key:
KNJDBYZZKAZQNG-UHFFFAOYSA-N
SMILES:
[O-][N+]([O-])=O.[O-][N+]([O-])=O.C[N+]1=C2C=CC=CC2=C(C2=CC=CC=C12)C1=C2C=CC=CC2=[N+](C)C2=CC=CC=C12
1
–
1
of
1
results
1
10,10'-Dimethyl-9,9'-biacridinium Dinitrate 97.0+%, TCI America™
CAS: 2315-97-1 Molecular Formula: C28H22N4O6 Molecular Weight (g/mol): 510.51 MDL Number: MFCD00011925 InChI Key: KNJDBYZZKAZQNG-UHFFFAOYSA-N Synonym: Lucigenin, Bis(N-methylacridinium) Nitrate PubChem CID: 129893650 IUPAC Name: 10,10'-dimethyl-[9,9'-biacridine]-10,10'-diium dinitrate SMILES: [O-][N+]([O-])=O.[O-][N+]([O-])=O.C[N+]1=C2C=CC=CC2=C(C2=CC=CC=C12)C1=C2C=CC=CC2=[N+](C)C2=CC=CC=C12