accessibility menu, dialog, popup

UserName

H3CNOO

CAS RN 2314-78-5

IUPAC Name: 1-ethylpyrrolidine-2,5-dione
Synonyms: 1-ethyl-pyrrolidine-2,5-dione
Molecular Weight (g/mol): 127.14
Molecular Formula: C6H9NO2
InChi Key: GHAZCVNUKKZTLG-UHFFFAOYSA-N
SMILES: CCN1C(=O)CCC1=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results