accessibility menu, dialog, popup

UserName

ClClClClClClKKRu

CAS RN 23013-82-3

IUPAC Name: λ⁴-ruthenium(4+) dipotassium hexachloride
Synonyms: λ⁴-ruthenium(4+) dipotassium hexachloride
Molecular Weight (g/mol): 391.97
Molecular Formula: Cl6K2Ru
InChi Key: XOLNQIIEFUNTQC-UHFFFAOYSA-H
SMILES: [Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[K+].[K+].[Ru+4]
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results