accessibility menu, dialog, popup

UserName

HNN

CAS RN 22961-45-1

IUPAC Name: N-phenylpyridin-4-amine
Synonyms: N-phenylpyridin-4-amine
Molecular Weight (g/mol): 170.22
Molecular Formula: C11H10N2
InChi Key: DKQSRQLSDPYGCJ-UHFFFAOYSA-N
SMILES: N(C1=CC=CC=C1)C1=CC=NC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results