accessibility menu, dialog, popup

UserName

CAS RN 2295-58-1

IUPAC Name: 1-(2,4,6-trihydroxyphenyl)propan-1-one
Synonyms: flopropione 夫洛丙酮 labrodax ecapron argobyl flopropionum phloropropiophenone labrodax supanate labroda cospanon
Molecular Weight (g/mol): 182.18
Molecular Formula: C9H10O4
InChi Key: PTHLEKANMPKYDB-UHFFFAOYSA-N
SMILES: CCC(=O)C1=C(O)C=C(O)C=C1O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results