CAS RN 2268-16-8
IUPAC Name:
2,4,5-trifluorophenol
Synonyms:
2,4,5-trifluorophenol
Molecular Weight (g/mol):
148.08
Molecular Formula:
C6H3F3O
InChi Key:
ODGMYCITQAIRCI-UHFFFAOYSA-N
SMILES:
OC1=CC(F)=C(F)C=C1F
1
–
1
of
1
results
1
2,4,5-Trifluorophenol, 94%
CAS: 2268-16-8 Molecular Formula: C6H3F3O Molecular Weight (g/mol): 148.084 MDL Number: MFCD00061215 InChI Key: ODGMYCITQAIRCI-UHFFFAOYSA-N Synonym: phenol, 2,4,5-trifluoro,phenol,2,4,5-trifluoro,2,4,5-tfifluorophenol,2,4,5-trifluoro-phenol,2,4,5-?trifluorophenol,acmc-1cne2,2,4,5-trifluorophenol,2,4,5-tris fluoranyl phenol,2,4,5-trifluorophenol 1gr PubChem CID: 123153 IUPAC Name: 2,4,5-trifluorophenol SMILES: C1=C(C(=CC(=C1F)F)F)O