accessibility menu, dialog, popup

UserName

CAS RN 22663-37-2

IUPAC Name: (4-chlorobenzenesulfonyl)urea
Synonyms: 4-chlorobenzenesulfonylurea
Molecular Weight (g/mol): 234.65
Molecular Formula: C7H7ClN2O3S
InChi Key: AZEPYUPSYWCRBG-UHFFFAOYSA-N
SMILES: NC(=O)NS(=O)(=O)C1=CC=C(Cl)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results