accessibility menu, dialog, popup

UserName

CAS RN 22513-32-2

ONHOH2ONa

CAS RN 22513-32-2

IUPAC Name: sodium hydrate (phenylformamido)olate
Synonyms: sodium hydrate benzohydroxamate
Molecular Weight (g/mol): 177.14
Molecular Formula: C7H8NNaO3
InChi Key: CTXNFHAZPJAZQN-UHFFFAOYSA-N
SMILES: O.[Na+].[O-]NC(=O)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results