accessibility menu, dialog, popup

UserName

OHFBr

CAS RN 222978-01-0

IUPAC Name: (4-bromo-3-fluorophenyl)methanol
Synonyms: (4-bromo-3-fluorophenyl)methanol
Molecular Weight (g/mol): 205.03
Molecular Formula: C7H6BrFO
InChi Key: VQDUFYPJCUBGQW-UHFFFAOYSA-N
SMILES: OCC1=CC(F)=C(Br)C=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)