CAS RN 22179-72-2
IUPAC Name:
4-fluorobenzene-1-carbothioamide
Synonyms:
4-fluorobenzenecarbothioamide
Molecular Weight (g/mol):
155.19
Molecular Formula:
C7H6FNS
InChi Key:
VQFOHZWOKJQOGO-UHFFFAOYSA-N
SMILES:
NC(=S)C1=CC=C(F)C=C1
1
–
1
of
1
results
1
Promotions Available
4-Fluorothiobenzamide, 97%
4-Fluorothiobenzamide, 97%
CAS: 22179-72-2 Molecular Formula: C7H6FNS Molecular Weight (g/mol): 155.19 MDL Number: MFCD01313285 InChI Key: VQFOHZWOKJQOGO-UHFFFAOYSA-N Synonym: 4-fluorothiobenzamide,4-fluorobenzothioamide,4-fluorobenzene-1-carbothioamide,4-fluoro-1-benzenecarbothioamide,4-fluoro-thiobenzamide,benzenecarbothioamide, 4-fluoro,amino 4-fluorophenyl methane-1-thione,p-fluorothiobenzamide,acmc-20amhq,4-fluoro thiobenzamide PubChem CID: 737223 IUPAC Name: 4-fluorobenzenecarbothioamide SMILES: NC(=S)C1=CC=C(F)C=C1