accessibility menu, dialog, popup

UserName

CAS RN 2216-33-3

H3CCH3CH3

CAS RN 2216-33-3

IUPAC Name: 3-methyloctane
Synonyms: 3-methyloctane
Molecular Weight (g/mol): 128.26
Molecular Formula: C9H20
InChi Key: SEEOMASXHIJCDV-UHFFFAOYNA-N
SMILES: CCCCCC(C)CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results