CAS RN 221302-75-6
IUPAC Name:
4-(2,6-dimethylphenoxy)benzene-1,2-dicarbonitrile
Synonyms:
4-(2,6-dimethylphenoxy)benzene-1,2-dicarbonitrile
Molecular Weight (g/mol):
248.29
Molecular Formula:
C16H12N2O
InChi Key:
LJUVUOSWARAIBZ-UHFFFAOYSA-N
SMILES:
CC1=CC=CC(C)=C1OC1=CC(C#N)=C(C=C1)C#N
1
–
1
of
1
results
1
4-(2',6'-Dimethylphenoxy)phthalonitrile 99.0+%, TCI America™
CAS: 221302-75-6 Molecular Formula: C16H12N2O Molecular Weight (g/mol): 248.285 MDL Number: MFCD00059064 InChI Key: LJUVUOSWARAIBZ-UHFFFAOYSA-N PubChem CID: 952073 IUPAC Name: 4-(2,6-dimethylphenoxy)benzene-1,2-dicarbonitrile SMILES: CC1=C(C(=CC=C1)C)OC2=CC(=C(C=C2)C#N)C#N