CAS RN 2207-01-4
IUPAC Name:
(1R,2S)-1,2-dimethylcyclohexane
Synonyms:
(1R,2S)-1,2-dimethylcyclohexane
Molecular Weight (g/mol):
112.22
Molecular Formula:
C8H16
InChi Key:
KVZJLSYJROEPSQ-OCAPTIKFSA-N
SMILES:
C[C@H]1CCCC[C@H]1C
1
–
1
of
1
results
1
cis-1,2-Dimethylcyclohexane 98.0+%, TCI America™
CAS: 2207-01-4 Molecular Formula: C8H16 Molecular Weight (g/mol): 112.22 MDL Number: MFCD00063621 InChI Key: KVZJLSYJROEPSQ-OCAPTIKFSA-N Synonym: cis-1,2-dimethylcyclohexane,1,cis-2-dimethylcyclohexane,1r,2s-1,2-dimethylcyclohexane,cyclohexane, 1,2-dimethyl-, cis,cyclohexane, 1,2-dimethyl-, 1r,2s,cyclohexane, 1,2-dimethyl-, 1r,2s-rel,cis-hexahydro-o-xylene,c-1,2-dimethylcyclohexane,1,2-cis-dimethyicyclohexane,1,2-dimethylcyclohexane, cis PubChem CID: 16628 IUPAC Name: (1S,2R)-1,2-dimethylcyclohexane SMILES: CC1CCCCC1C