CAS RN 220227-93-0
IUPAC Name:
1-[2-(trifluoromethoxy)phenyl]ethan-1-one
Synonyms:
1-[2-(trifluoromethoxy)phenyl]ethanone
Molecular Weight (g/mol):
204.15
Molecular Formula:
C9H7F3O2
InChi Key:
XTXSTESGCXKUIH-UHFFFAOYSA-N
SMILES:
CC(=O)C1=CC=CC=C1OC(F)(F)F
1
–
1
of
1
results
1
2'-(Trifluoromethoxy)acetophenone 98.0+%, TCI America™
CAS: 220227-93-0 Molecular Formula: C9H7F3O2 Molecular Weight (g/mol): 204.148 MDL Number: MFCD00236320 InChI Key: XTXSTESGCXKUIH-UHFFFAOYSA-N Synonym: 2'-trifluoromethoxy acetophenone,1-2-trifluoromethoxy phenyl ethanone,1-2-trifluoromethoxy phenyl ethan-1-one,1-acetyl-2-trifluoromethoxy benzene,pubchem4295,2-trifluoromethoxy acetophenone98,1-2-trifluoromethoxyphenyl ethanone,2'-trifluoromethoxy acetophenone98 PubChem CID: 2777176 IUPAC Name: 1-[2-(trifluoromethoxy)phenyl]ethanone SMILES: CC(=O)C1=CC=CC=C1OC(F)(F)F