CAS RN 2201-23-2
IUPAC Name:
1-phenylcyclohexane-1-carbonitrile
Synonyms:
1-phenylcyclohexane-1-carbonitrile
Molecular Weight (g/mol):
185.27
Molecular Formula:
C13H15N
InChi Key:
AUXIEQKHXAYAHG-UHFFFAOYSA-N
SMILES:
N#CC1(CCCCC1)C1=CC=CC=C1
1
–
1
of
1
results
1
1-Phenylcyclohexanecarbonitrile 97.0+%, TCI America™
CAS: 2201-23-2 Molecular Formula: C13H15N Molecular Weight (g/mol): 185.27 MDL Number: MFCD00021281 InChI Key: AUXIEQKHXAYAHG-UHFFFAOYSA-N Synonym: 1-phenylcyclohexanecarbonitrile,1-phenyl-1-cyclohexanecarbonitrile,cyclohexanecarbonitrile, 1-phenyl,cyanophenylcyclohexane,1-cyano-1-phenylcyclohexane,1-phenyl-cyclohexanecarbonitrile,1-phenylcyclohexane carbonitrile,1-phenyl-1-cyclohexane-carbonitrile PubChem CID: 75148 IUPAC Name: 1-phenylcyclohexane-1-carbonitrile SMILES: N#CC1(CCCCC1)C1=CC=CC=C1