accessibility menu, dialog, popup

UserName

CAS RN 219968-28-2

IUPAC Name: benzyl 3-{[4,5-bis(benzyloxy)-6-[(benzyloxy)methyl]-3-acetamidooxan-2-yl]carbamoyl}-2-{[(tert-butoxy)carbonyl]amino}propanoate
Synonyms: benzyl 3-{[4,5-bis(benzyloxy)-6-[(benzyloxy)methyl]-3-acetamidooxan-2-yl]carbamoyl}-2-[(tert-butoxycarbonyl)amino]propanoate
Molecular Weight (g/mol): 795.93
Molecular Formula: C45H53N3O10
InChi Key: CRNGSGNVFXQOBD-UHFFFAOYNA-N
SMILES: CC(=O)NC1C(NC(=O)CC(NC(=O)OC(C)(C)C)C(=O)OCC2=CC=CC=C2)OC(COCC2=CC=CC=C2)C(OCC2=CC=CC=C2)C1OCC1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results