accessibility menu, dialog, popup

UserName

H3C(S)NHOOHO

CAS RN 21752-36-3

IUPAC Name: 2-{[(1S)-1-phenylethyl]carbamoyl}benzoic acid
Synonyms: 2-{[(1S)-1-phenylethyl]carbamoyl}benzoic acid
Molecular Weight (g/mol): 269.3
Molecular Formula: C16H15NO3
InChi Key: VCFKXWGKKDZMPO-NSHDSACASA-N
SMILES: C[C@H](NC(=O)C1=CC=CC=C1C(O)=O)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results