accessibility menu, dialog, popup

UserName

HOOClBr

CAS RN 21739-93-5

IUPAC Name: 2-bromo-5-chlorobenzoic acid
Synonyms: 2-bromo-5-chlorobenzoic acid
Molecular Weight (g/mol): 235.46
Molecular Formula: C7H4BrClO2
InChi Key: RBCPJQQJBAQSOU-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(Cl)=CC=C1Br
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)