accessibility menu, dialog, popup

UserName

ClClH3CHN(R)(S)HO(S)HNCH3(R)O(R)OH(S)O(R)CH3OO(R)(S)OHHH

CAS RN 21736-83-4

IUPAC Name: dihydrogen (1R,3S,5R,8R,10R,11S,12S,13R,14S)-8,12,14-trihydroxy-5-methyl-11,13-bis(methylamino)-2,4,9-trioxatricyclo[8.4.0.0³,⁸]tetradecan-7-one dichloride
Synonyms: dihydrogen spectinomycin dichloride
Molecular Weight (g/mol): 405.27
Molecular Formula: C14H26Cl2N2O7
InChi Key: FOGDPGQSHLLYIK-XYQGXRRISA-N
SMILES: [H+].[H+].[Cl-].[Cl-].CN[C@@H]1[C@H](O)[C@H](NC)[C@H]2O[C@]3(O)[C@@H](O[C@H](C)CC3=O)O[C@@H]2[C@H]1O
Color 
  • (1)
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results