accessibility menu, dialog, popup

UserName

OO

CAS RN 2170-13-0

IUPAC Name: [1,1'-biphenyl]-4-yl benzoate
Synonyms: [1,1'-biphenyl]-4-yl benzoate
Molecular Weight (g/mol): 274.32
Molecular Formula: C19H14O2
InChi Key: CINHWMYRCOGYIX-UHFFFAOYSA-N
SMILES: O=C(OC1=CC=C(C=C1)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results