accessibility menu, dialog, popup

UserName

BrOBr

CAS RN 216312-73-1

IUPAC Name: 3,6-dibromo-9H-fluoren-9-one
Synonyms: 3,6-dibromofluoren-9-one
Molecular Weight (g/mol): 338
Molecular Formula: C13H6Br2O
InChi Key: XXZOHYHKWDLSFS-UHFFFAOYSA-N
SMILES: BrC1=CC2=C(C=C1)C(=O)C1=C2C=C(Br)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results