accessibility menu, dialog, popup

UserName

CAS RN 2147-83-3

IUPAC Name: 4-(2-oxo-2,3-dihydro-1H-1,3-benzodiazol-1-yl)-1,2,3,6-tetrahydropyridin-1-ium
Synonyms: 4-(2-oxo-3H-1,3-benzodiazol-1-yl)-1,2,3,6-tetrahydropyridin-1-ium
Molecular Weight (g/mol): 216.26
Molecular Formula: C12H14N3O
InChi Key: YFEOSTXFQCDCAR-UHFFFAOYSA-O
SMILES: O=C1NC2=CC=CC=C2N1C1=CC[NH2+]CC1
Percent Purity 
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)
Quantity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)