CAS RN 21436-98-6
CAS RN 21436-98-6
IUPAC Name:
hydrogen 2,6-dimethylaniline chloride
Synonyms:
2,6-xylidine, hydrochloride
Molecular Weight (g/mol):
157.64
Molecular Formula:
C8H12ClN
InChi Key:
QNWMFJDRMZWZNN-UHFFFAOYSA-N
SMILES:
[H+].[Cl-].CC1=CC=CC(C)=C1N
1
–
1
of
1
results
1
2,6-Dimethylaniline Hydrochloride 98.0+%, TCI America™
CAS: 21436-98-6 Molecular Formula: C8H12ClN Molecular Weight (g/mol): 157.641 MDL Number: MFCD00060213 InChI Key: QNWMFJDRMZWZNN-UHFFFAOYSA-N Synonym: 2,6-Xylidine Hydrochloride PubChem CID: 88899 IUPAC Name: 2,6-dimethylaniline;hydrochloride SMILES: CC1=C(C(=CC=C1)C)N.Cl